Learn More
Norell™ Adapter Optimizer Insert Bruker for 5mm spinner turbine 4mm Norell
Product Code. 12832326
Product Code. 12832326 Supplier Norell™ Supplier No. 5X4INSB
Only null left
Add to basket
Add to basket
Precision adapter holds 3mm, 4mm or 4.25mm NMR tube in 5mm spinner turbine.
Available for Bruker and Varian spectrometers. Does not include spinner turbine.