Learn More
Norell™ Adapter Optimizer Insert Bruker for 5mm spinner turbine 4.25mm Norell
Product Code. 12852326
Product Code. 12852326 Supplier Norell™ Supplier No. 5X4.25INSB
Only null left
Add to basket
Add to basket
Precision adapter holds 3mm, 4mm or 4.25mm NMR tube in 5mm spinner turbine.
Available for Bruker and Varian spectrometers. Does not include spinner turbine.