missing translation for 'onlineSavingsMsg'
Learn More

Berthold Technologies Adapter For Microcentrifuge Tubes

Product Code. 31777304 Shop All Berthold Technologies Products
Change view
Click to view available options
Includes:
Adapter (1 Pc.)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
31777304 Adapter (1 Pc.) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31777304 Supplier Berthold Technologies Supplier No. 57064
Estimated Shipment: 02-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For the Berthold Lumat LB 9510 tube luminometer

  • Adapters for microcentrifuge tubes are already included with the Lumat, but they can be purchased separately if needed.

Specifications

Type Adapter
Color Gray
For Use With (Application) Luminescence Assays
For Use With (Equipment) Lumat LB 9510 Tube Luminometer
Includes Adapter (1 Pc.)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.