Learn More
Cole-Parmer™ Stuart™ SHM/ADAPT Adaptor for Disposable Homogenizer Probes
Make the most of your homogenizer
Brand: Cole-Parmer™ 04720-08
Code : 04
Additional Details : Weight : 1.00000kg
Description
- Probes available in different sizes and materials
- Stand for benchtop use
- Parts and adapters to expand capabilities
- Compatible with SHM1 and SHM3 Adapts disposable probes to homogenizer
Specifications
Adapter | |
SHM | |
1 Ea. |