missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mettler Toledo™ Adapter
Brand: Mettler Toledo™ 51340024
13.01 GBP valid until 2024-05-31
Use promo code "21700" to get your promotional price.
Code : 59
Additional Details : Weight : 0.01000kg
Specifications
Adapter | |
12mm Electrode |