missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ Adaptaquin

Description
Adaptaquin is a HIF-prolyl hydroxylase-2 (PHD2) inhibitor. Blocks glutamate induced ROS production in HT-22 cells, independent of MnSOD. Exhibits neuroprotective effects and enhances functional recovery in rodent intracerebral hemorrhage models, via inhibition of ATF4 dependent genes. Brain penetrant. Also antioxidant.
Specifications
Specifications
| Chemical Name or Material | 7-[(4-Chlorophenyl)[(3-hydroxy-2-pyridinyl)amino]methyl]-8-quinolinol |
| CAS | 385786-48-1 |
| Quantity | 50 mg |
| Target | Hydrolase Inhibitors |
| Molecular Formula | C21H16ClN3O2 |
| Formula Weight | 377.82 |
| Purity | >97% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?