missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Adapalene USP Related Compound B, Mikromol™
Description
Adapalene USP Related Compound B
Specifications
Specifications
| Chemical Name or Material | Methyl 6-[3-(1-Adamantyl)-4-methoxyphenyl]-2-naphthoate (Adapalene Methyl Ester) |
| CAS | 106685-41-0 |
| Analyte or Component Names | Methyl 6-[3-(1-Adamantyl)-4-methoxyphenyl]-2-naphthoate (Adapalene Methyl Ester) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C29 H30 O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 2-Naphthalenecarboxylic acid, 6-(4-methoxy-3-tricyclo[3.3.1.13,7]dec-1-ylphenyl)-, methyl ester, Adapalene Related Compound B |
| InChI Formula | InChI=1S/C29H30O3/c1-31-27-8-7-24(14-26(27)29-15-18-9-19(16-29)11-20(10-18)17-29)22-3-4-23-13-25(28(30)32-2)6-5-21(23)12-22/h3-8,12-14,18-20H,9-11,15-17H2,1-2H3 |
| SMILES | COC(=O)c1ccc2cc(ccc2c1)c3ccc(OC)c(c3)C45CC6CC(CC(C6)C4)C5 |
| IUPAC Name | methyl 6-[3-(1-adamantyl)-4-methoxyphenyl]naphthalene-2-carboxylate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?