missing translation for 'onlineSavingsMsg'
Learn More

Adapalene, TRC

Product Code. 30317310
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30317310 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30317310 Supplier Toronto Research Chemicals Supplier No. TRCA22500010MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Adapalene
CAS 106685-40-9
Molecular Formula C28 H28 O3
Quantity 10 mg
Synonym Adapalene, 6-(4-Methoxy-3-tricyclo[3.3.1.1(3,7)]dec-1-ylphenyl)naphthalene-2-carboxylic acid, 6-(3-(Adamantan-1-yl)-4-methoxyphenyl)-2-naphthoic acid, 6-[3-(1-Adamantyl)-4-methoxyphenyl]-2-naphthalenecarboxylic acid, 6-[3-(1-Adamantyl)-4-methoxyphenyl]-2-naphthoic acid, Aclene, Aclene Gel, CD 271, CD 271 (pharmaceutical), Differin, Differin Gel
InChI Formula InChI=1S/C28H28O3/c1-31-26-7-6-23(21-2-3-22-12-24(27(29)30)5-4-20(22)11-21)13-25(26)28-14-17-8-18(15-28)10-19(9-17)16-28/h2-7,11-13,17-19H,8-10,14-16H2,1H3,(H,29,30)
SMILES COc1ccc(cc1C23CC4CC(CC(C4)C2)C3)c5ccc6cc(ccc6c5)C(=O)O
IUPAC Name 6-[3-(1-adamantyl)-4-methoxyphenyl]naphthalene-2-carboxylic acid
Molecular Weight (g/mol) 412.52
Formula Weight 412.2038
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.