missing translation for 'onlineSavingsMsg'
Learn More

Adapalene Methyl Ester, TRC

Product Code. 30132602
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
100mg
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132602 250 mg 250mg
30007999 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30132602 Supplier Toronto Research Chemicals Supplier No. TRCA225010250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106685-41-0
Molecular Formula C29 H30 O3
Molecular Weight (g/mol) 426.55
Synonym 2-Naphthalenecarboxylic acid, 6-(4-methoxy-3-tricyclo[3.3.1.13,7]dec-1-ylphenyl)-, methyl ester
IUPAC Name methyl 6-[3-(1-adamantyl)-4-methoxyphenyl]naphthalene-2-carboxylate
SMILES COC(=O)c1ccc2cc(ccc2c1)c3ccc(OC)c(c3)C45CC6CC(CC(C6)C4)C5

Specifications

Analyte or Component Names Methyl 6-[3-(1-Adamantyl)-4-methoxyphenyl]-2-naphthoate (Adapalene Methyl Ester)
Shipping Condition Room Temperature
InChI Formula InChI=1S/C29H30O3/c1-31-27-8-7-24(14-26(27)29-15-18-9-19(16-29)11-20(10-18)17-29)22-3-4-23-13-25(28(30)32-2)6-5-21(23)12-22/h3-8,12-14,18-20H,9-11,15-17H2,1-2H3
Solution Type Neat
Industry Type Pharmaceutical
Quantity 250 mg
Formula Weight 426.2195
Percent Purity >95%
Chemical Name or Material Methyl 6-[3-(1-Adamantyl)-4-methoxyphenyl]-2-naphthoate (Adapalene Methyl Ester)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.