missing translation for 'onlineSavingsMsg'
Learn More

Adapalene, MedChemExpress

Product Code. p-200101839
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
This item is not returnable. View return policy

Product Code. 16442006

Brand: Medchem Express HYB0091

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Adapalene (CD271), a third-generation synthetic retinoid, is widely used for the research of acne. Adapalene is a potent RAR agonist, with AC50s of 2.3 nM, 9.3 nM, and 22 nM for RARβ, RARγ, RARα, respectively. Adapalene also inhibits the enzymatic activity of GOT1 in a non-competitive manner. Adapalene exhibits anti-tumor activity.

Adapalene (CD271), a third-generation synthetic retinoid, is widely used for the research of acne. Adapalene is a potent RAR agonist, with AC50s of 2.3 nM, 9.3 nM, and 22 nM for RARβ, RARγ, RARα, respectively. Adapalene also inhibits the enzymatic activity of GOT1 in a non-competitive manner. Adapalene exhibits anti-tumor activity.

Specifications

Chemical Name or Material Adapalene
CAS 106685-40-9
Color White
Purity Grade Notes Research
Molecular Formula C28H28O3
Quantity 500 mg
Synonym CD271
Solubility Information DMSO : 20 mg/mL (48.48 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES COC1=C(C23CC4CC(C3)CC(C2)C4)C=C(C5=CC6=C(C=C(C(O)=O)C=C6)C=C5)C=C1
Molecular Weight (g/mol) 412.52
Formula Weight 412.52
Percent Purity 97.0%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.