missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ADAMTSL5 Polyclonal Antibody

Product Code. 12839082
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12839082 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12839082 Supplier Invitrogen™ Supplier No. PA520934

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chicken Polyclonal Antibody

A suggested positive control is human skeletal muscle tissue lysate. PA5-20934 can be used with blocking peptide PEP-1048.

ADAMTSL5 belongs to a large superfamily containing 20 ADAMTS proteases and at least eight ADAMTS-like proteins. ADAMTS proteases are secreted enzymes with a conserved organization that includes a metalloprotease domain and an ancillary domain containing one or more thrombospondin type 1 repeats (TSR). The ADAMTS-like subfamily comprises proteins homologous to the ADAMTS ancillary domains but lacking the protease domain and hence lacking catalytic activity. The exact role of ADAMTSL5 is currently unknown, but other ADAMTS-like proteins appear to have regulatory roles in the extracellular matrix.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAMTSL5
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene ADAMTSL5
Gene Accession No. Q6ZMM2
Gene Alias 2010109H09Rik; a disintegrin and metalloproteinase; ADAM; ADAMs; ADAMTS like 5; ADAMTSL5; ADAMTSL-5; ADAMTS-like 5; ADAMTS-like protein 5; metalloendopeptidases; Thrombospondin type-1 domain-containing protein 6; thrombospondin, type I domain containing 6; thrombospondin, type I, domain containing 6; THSD6
Gene Symbols ADAMTSL5
Host Species Chicken
Immunogen A 17 amino acid peptide near the carboxy terminus of human ADAMTSL5.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 314626, 339366, 66548
Target Species Human, Mouse, Rat
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.