missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAMTSL5 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NBP2-41120AF750
This item is not returnable.
View return policy
Description
ADAMTSL5 Polyclonal antibody specifically detects ADAMTSL5 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| ADAMTSL5 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Chicken | |
| Peptide affinity purified | |
| RUO | |
| 339366 | |
| Store at 4°C in the dark. | |
| IgY |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 750 | |
| ADAMTSL-5, ADAMTS-like 5, ADAMTS-like protein 5, thrombospondin type-1 domain-containing protein 6, thrombospondin, type I, domain containing 6, THSD6 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the carboxy terminus of human ADAMTSL5. The immunogen is located within the last 50 amino acids of ADAMTSL5. Amino Acid Squence: YARPWSPAEDSRIRLTA | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction