missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ADAMTSL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561057
This item is not returnable.
View return policy
Description
Immunogen sequence: PWSECTKTCG VGVRMRDVKC YQGTDIVRGC DPLVKPVGRQ ACDLQPCPTE PPDDSCQDQP GTNCALAIKV NLCGHWYYSK ACC Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
ADAMTSL2, or A Disintegrin And Metalloproteinase with Thrombospondin Motifs Like 2, is a gene that codes for a secreted glycoprotein involved in the regulation of extracellular matrix (ECM) and associated proteases. The gene, initially identified by the Kazusa DNA Research Institute, plays a crucial role in various biological processes including ECM remodeling. Mutations or alterations in ADAMTSL2 are implicated in multiple diseases, including genetic disorders like geleophysic dysplasia and cancer. Its expression levels and genetic variants have been correlated with cancer progression, immune cell infiltration in tumors, and other pathological conditions like non-alcoholic steatohepatitis (NASH). ADAMTSL2 is a potential biomarker for disease prognosis and therapeutic targeting due to its significant role in ECM dynamics and disease pathology.
Specifications
| ADAMTSL2 | |
| Polyclonal | |
| Unconjugated | |
| ADAMTSL2 | |
| a disintegrin and metalloproteinase; A930008K15Rik; ADAM; ADAMs; ADAMTS like 2; Adamtsl2; ADAMTSL-2; ADAMTS-like 2; ADAMTS-like protein 2; GPHYSD1; Kiaa0605; metalloendopeptidases; RGD1305459; Tcp1; TCP-1; TSP1-repeat-containing protein 1; TSP1-repeats containing protein; TSP1-repeats-containing protein 1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 9719 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86TH1 | |
| ADAMTSL2 | |
| Recombinant protein corresponding to Human ADAMTSL2. Recombinant protein control fragment (Product #RP-99616). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction