missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ADAMTSL1 Polyclonal Antibody

Description
Immunogen sequence: QVPTQLEDIR ALLAATGPNL PSVLTSPLGT QLVLDPGNSA LLGCPIKGHP VPNITWFHGG QPIVTATGLT HHILAAGQIL QVANLSGGSQ GEFSCLAQNE AGVLMQKASL VIQDYWWSVD RLATCS Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 89%.
Punctin (ADAMTSL-1) is a secreted molecule, expressed in adult skeletal muscle, resembling members of the ADAMTS family of proteases, containing four thrombospondin type I repeats. Human punctin cleaves aggrecan and is thereby classified as an aggrecanase. Like the ADAMTS proteases, each domain in punctin has an even number of cysteine residues suggesting that each domain may have internal disulfide bonds and that punctin consists of a series of independently folded and disulfide-bonded domains.
Specifications
Specifications
| Antigen | ADAMTSL1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ADAMTSL1 |
| Gene Accession No. | Q8N6G6 |
| Gene Alias | 5930437A14Rik; 6720426B09Rik; a disintegrin and metalloproteinase; ADAM; ADAMs; ADAMTS like 1; ADAM-TS related protein 1; Adamtsl1; ADAMTSL-1; ADAMTS-like 1; ADAMTS-like protein 1; ADAMTSR1; C9orf94; DKFZp686L03130; FLJ35283; FLJ41032; FLJ46891; metalloendopeptidases; MGC118803; MGC118805; MGC40193; PUNCTIN; Punctin-1; RP11-220B22.2; UNQ528/PRO1071 |
| Gene Symbols | ADAMTSL1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?