Learn More
Invitrogen™ ADAMTS8 Polyclonal Antibody

Sheep Polyclonal Antibody
Marque: Invitrogen™ PA547031
Description
In direct ELISAs, approximately 5% cross-reactivity with recombinant human (rh) ADAMTS1 is observed and less than 1% cross-reactivity with rhADAMTS1, rhADAMTS3, rhADAMTS4, rhADAMTS5, rhADAMTS10, rhADAMTS12, rhADAMTS13, and rhADAMTS16 is observed. Reconstitute in sterile PBS to a final concentration of 0.2 mg/mL.
This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs) protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The enzyme encoded by this gene contains two C-terminal TS motifs, and disrupts angiogenesis in vivo. A number of disorders have been mapped in the vicinity of this gene, most notably lung neoplasms.
Spécification
| ADAMTS8 | |
| Polyclonal | |
| Unconjugated | |
| ADAMTS8 | |
| a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 8; a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 8; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 8; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 8; ADAM metallopeptidase with thrombospondin type 1 motif, 8; ADAMs; ADAM-TS 8; ADAMTS8; ADAM-TS8; ADAMTS-8; ADAMTS-8 precursor (A disintegrin and metalloproteinase with thrombospondin motifs 8) (ADAM-TS 8) (ADAM-TS8) (METH-2); EC 3.4.24.14; EC 3.4.24.82; FLJ41712; metalloendopeptidases; METH2; METH-2; METH2ADAM-TS METH-8; METH-8 | |
| Sheep | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11095 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q9UP79 | |
| ADAMTS8 | |
| CHO-derived recombinant human ADAMTS8 Pro29-Arg691 (Gly35Arg, Gly431Ala, Val526Ala). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.