missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAMTS8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA564274
This item is not returnable.
View return policy
Description
Immunogen sequence: SIATLERLQS FRPLPEPLTV QLLTVPGEVF PPKVKYTFFV PNDVDFSMQS SKERATTNII QPLLHAQWVL Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 83%.
This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs) protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The enzyme encoded by this gene contains two C-terminal TS motifs, and disrupts angiogenesis in vivo. A number of disorders have been mapped in the vicinity of this gene, most notably lung neoplasms.
Specifications
| ADAMTS8 | |
| Polyclonal | |
| Unconjugated | |
| ADAMTS8 | |
| a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 8; a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 8; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 8; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 8; ADAM metallopeptidase with thrombospondin type 1 motif, 8; ADAMs; ADAM-TS 8; ADAMTS8; ADAM-TS8; ADAMTS-8; ADAMTS-8 precursor (A disintegrin and metalloproteinase with thrombospondin motifs 8) (ADAM-TS 8) (ADAM-TS8) (METH-2); EC 3.4.24.14; EC 3.4.24.82; FLJ41712; metalloendopeptidases; METH2; METH-2; METH2ADAM-TS METH-8; METH-8 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11095 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UP79 | |
| ADAMTS8 | |
| Recombinant protein corresponding to Human ADAMTS8 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction