missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAMTS8 Antibody (OTI2H5), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71677PCP
This item is not returnable.
View return policy
Description
ADAMTS8 Monoclonal antibody specifically detects ADAMTS8 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| ADAMTS8 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunocytochemistry | |
| OTI2H5 | |
| Western Blot, Immunocytochemistry/ Immunofluorescence | |
| a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif, 8, ADAM metallopeptidase with thrombospondin type 1 motif, 8, ADAMTS-8, ADAM-TS8A disintegrin and metalloproteinase with thrombospondin motifs 8, EC 3.4.24, EC 3.4.24.-, EC 3.4.24.14, EC 3.4.24.82, FLJ41712, METH-2, METH2ADAM-TS 8, METH-8 | |
| Full length human recombinant protein of human ADAMTS8(NP_008968) produced in HEK293T cell. | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 11095 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction