missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ ADAMTS8 Antibody (5A3), Novus Biologicals™

Product Code. 18347299 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18347299 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18347299 Supplier Novus Biologicals™ Supplier No. H00011095M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ADAMTS8 Monoclonal antibody specifically detects ADAMTS8 in Human samples. It is validated for ELISA,Western Blot,Sandwich ELISA,Protein Array
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAMTS8
Applications ELISA, Western Blot, Sandwich ELISA, Protein Array
Classification Monoclonal
Clone 5A3
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA, Protein Array
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_008968
Gene Alias a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif, 8, ADAM metallopeptidase with thrombospondin type 1 motif, 8, ADAMTS-8, ADAM-TS8A disintegrin and metalloproteinase with thrombospondin motifs 8, EC 3.4.24, EC 3.4.24.-, EC 3.4.24.14, EC 3.4.24.82, FLJ41712, METH-2, METH2ADAM-TS 8, METH-8
Host Species Mouse
Immunogen ADAMTS8 (NP_008968, 781 a.a. ∽ 890 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. RPLPEPLTVQLLTVPGEVFPPKVKYTFFVPNDVDFSMQSSKERATTNIIQPLLHAQWVLGDWSECSSTCGAGWQRRTVECRDPSGQASATCNKALKPEDAKPCESQLCPL
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 11095
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.