Learn More
ADAMTS7 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26836-1-AP-20UL
Description
The ADAMTS7 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human ADAMTS7. This antibody recognizes human, mouse antigen. The ADAMTS7 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| ADAMTS7 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 11173 | |
| IgG |
| Western Blot, Immunohistochemistry, ELISA | |
| 200 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse |