missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ADAMTS7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560972
This item is not returnable.
View return policy
Description
Immunogen sequence: LGRAHIRAHT PACHLLGEVQ DSELEGGLAA ISACDGLKGV FLLSN Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 71%.
ADAMTS (a disintegrin and metalloproteinase domain with thrombospondin type-1 modules) is a family of zinc-dependent proteases that are implicated in a variety of normal and pathological conditions, including arthritis and cancer. ADAMTS protein family members contain an N-terminal propeptide domain, a metalloproteinase domain, a disintegrin-like domain and a C-terminus that contains a varying number of thrombospondin type-1 (TSP-1) motifs. ADAMTS genes are primarily expressed in fetal tissues, including lung, kidney and liver. ADAMTS-7 (ADAM metallopeptidase with thrombospondin type 1 motif, 7), also known as COMPase, is a 1,686 amino acid protein that exists as two alternatively spliced isoforms. Encoded by a gene that maps to human chromosome 15q25.1, ADAMTS-7 contains eight TSP-1 motifs and binds one zinc ion per subunit. ADAMTS-7 is expressed in heart, brain, placenta, lung, liver, skeletal muscle, kidney and pancreas. ADAMTS-7 is also located in meniscus, bone, tendon, cartilage, synovium, fat and ligaments, and is up-regulated in articular cartilage and synovium in arthritis patients. ADAMTS-7 functions as a metalloprotease and may play a role in the degradation of COMP. ADAMTS-7 is pH dependent, with optimum pH between 7.5 and 9.5.
Specifications
| ADAMTS7 | |
| Polyclonal | |
| Unconjugated | |
| ADAMTS7 | |
| a disintegrin and metalloprotease with thrombospondin motifs-7 preproprotein; a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 7; a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 7; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 7; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 7; ADAM metallopeptidase with thrombospondin type 1 motif, 7; ADAMs; ADAM-TS 7; ADAMTS7; ADAM-TS7; ADAMTS-7; ADAMTS7B; COMPase; metalloendopeptidases; metalloprotease | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 11173 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UKP4 | |
| ADAMTS7 | |
| Recombinant protein corresponding to Human ADAMTS7. Recombinant protein control fragment (Product #RP-102975). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction