missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ADAMTS5 Polyclonal Antibody

Description
Recommended positive controls: HCT116, MCF-7, JC. Predicted reactivity: Mouse (100%), Rat (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ADAMTS5, also known as Implantin or Aggrecanase 2, is a member of the larger family of ADAMs (A Disintegrin And Metalloproteinase) metalloproteinases containing thrombospondin (TS) repeats. ADAMTS5 (A Disintegrin And Metalloproteinase with Thrombospondin-5 motif) was first described as ADAMTS5, a protein elevated in mice during the peri-implantion period. At the same time, another group identified Aggrecanase 11, a protein elevated in arthritic synovium. The name was later changed to ADAMTS5. ADAMTS5 is expressed in human and mouse. It has been found in heart, lung, cervix, uterus, ovary, brain, cartilage, and numerous other tissues, as well as chondroblastoma cell lines.
Specifications
Specifications
| Antigen | ADAMTS5 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.01% thimerosal; pH 7 |
| Gene | ADAMTS5 |
| Gene Accession No. | Q9R001, Q9UNA0 |
| Gene Alias | 9530092O11Rik; a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 11; A disintegrin and metalloproteinase with thrombospondin motifs 5; a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 5 (aggrecanase-2); a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 5; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 5 (aggrecanase-2); ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 5; ADAM metallopeptidase with thrombospondin type 1 motif, 5; ADAMs; ADAM-TS 11; ADAM-TS 5; ADAMTS family member; ADAMTS1; ADAMTS11; ADAMTS-11; ADAMTS5; ADAM-TS5; ADAMTS-5; ADMP2; ADMP-2; Aggrecanase 2; Aggrecanase-2; AI481094; ASMP-2; Implantin; metalloendopeptidases |
| Gene Symbols | ADAMTS5 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?