Learn More
ADAMTS4 Rabbit, anti-Human, Clone: 4F9, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand: Proteintech 82744-2-RR-20UL
Description
The ADAMTS4 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human ADAMTS4. This antibody recognizes human, mouse, rat antigen. The ADAMTS4 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| ADAMTS4 | |
| 4F9 | |
| ADAM TS 4, ADAM TS4, ADAMTS 2, ADAMTS 4, ADAMTS4, ADMP 1, Aggrecanase 1, KIAA0688 | |
| 20 μL | |
| 9507 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG |