Learn More
Invitrogen™ ADAMTS4 Polyclonal Antibody

Description
PA1-1749A detects ADAMTS4 from porcine, bovine, mouse and human samples. PA1-1749A is specific for ADAMTS4 and shows no cross-reactivity with other ADAMTS family members. PA1-1749A has been successfully used in Western blot procedures. By Western blot, this antibody detects ADAMTS4 at ~68, ~53 and 30 kDa protein representing ADAMTS4 and its proteolytic fragments. Depending on the extent of C-terminal processing, several smaller bands may also be seen using this antibody. PA1-1749A immunogen is a synthetic peptide corresponding to residues C V(394) M A H V D P E E P(403) of mouse ADAMTS4. PA1-1749A immunizing peptide (Cat. # PEP-234) is available for use in neutralization and control procedures. This polyclonal antibody has been referred to as: JSCVMA. PA1-1749A has been used in combination with PA1-1750 for detection of all ADAMTS4 processed forms.
Specifications
Specifications
| Antigen | ADAMTS4 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA and 0.05% sodium azide |
| Gene | ADAMTS4 |
| Gene Accession No. | O75173, Q8BNJ2, Q9TT93 |
| Gene Alias | 3830423K05; a disintegrin and metallopeptidase with thrombospondin motifs 4; a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 4; a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 4; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 4; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 4; ADAM metallopeptidase with thrombospondin type 1 motif, 4; ADAMs; ADAM-TS 4; ADAMTS family member; ADAMTS-2; Adamts4; ADAM-TS4; ADAMTS-4; ADMP-1; Aggrecanase; Aggrecanase 1; aggrecanase-1; disintegrin and metalloproteinase with thrombospondin motifs 2; KIAA0688; metalloendopeptidases; metalloproteinase; mKIAA0688; UNQ769/PRO1563 |
| Gene Symbols | ADAMTS4 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.