missing translation for 'onlineSavingsMsg'
Learn More

ADAMTS4 Antibody, DyLight 550, Novus Biologicals™

Product Code. 18392943 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18392943 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18392943 Supplier Novus Biologicals Supplier No. NBP299438R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ADAMTS4 Polyclonal specifically detects ADAMTS4 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAMTS4
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 550
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif, 4, ADAM metallopeptidase with thrombospondin type 1 motif, 4, ADAM-TS 4, ADAMTS-2, ADAM-TS4, ADAMTS-4, ADMP-1EC 3.4.24.82, aggrecanase-1, EC 3.4.24, KIAA0688A disintegrin and metalloproteinase with thrombospondin motifs 4
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of mouse ADAMTS4.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, GPCR, Neuroscience, Vision
Primary or Secondary Primary
Gene ID (Entrez) 9507
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.