missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAMTS3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £470.00
Specifications
| Antigen | ADAMTS3 |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18426921
|
Novus Biologicals
NBP2-30495-25ul |
25ul |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18101174
|
Novus Biologicals
NBP2-30495 |
0.1 mL |
£470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ADAMTS3 Polyclonal specifically detects ADAMTS3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| ADAMTS3 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| A disintegrin and metalloproteinase with thrombospondin motifs 3, a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif, 3, ADAM metallopeptidase with thrombospondin type 1 motif, 3, ADAM-TS 3, ADAM-TS3, ADAMTS-3, ADAMTS-4, EC 3.4.24, EC 3.4.24.-, EC 3.4.24.14, KIAA0366PC II-NP, Procollagen II amino propeptide-processing enzyme, Procollagen II N-proteinase, zinc metalloendopeptidase | |
| ADAMTS3 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| O15072 | |
| 9508 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: QAGNEEMVQIDLPIKRYREYELVTPVSTNLEGRYLSHTLSASHKKRSARDVSSNPEQLFFNITAFGKDFHLRLKPNTQL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title