missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAMTS2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£161.00 - £368.00
Specifications
| Antigen | ADAMTS2 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18619910
|
Novus Biologicals
NBP2-92008-0.02ml |
0.02 mL |
£161.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18698220
|
Novus Biologicals
NBP2-92008-0.1ml |
0.1 mL |
£368.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ADAMTS2 Polyclonal antibody specifically detects ADAMTS2 in Human, Rat samples. It is validated for Western BlotSpecifications
| ADAMTS2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Cancer, Extracellular Matrix | |
| PBS with 50% glycerol, pH7.3. | |
| 9509 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| A disintegrin and metalloproteinase with thrombospondin motifs 2, a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif, 2, ADAM metallopeptidase with thrombospondin type 1 motif, 2, ADAM-TS 2, ADAMTS-2, ADAM-TS2EC 3.4.24.14, ADAMTS-3, EC 3.4.24, NPIDKFZp686F12218, PC I-NP, PCI-NP, PCINPhPCPNI, PCPNI, pNPI, Procollagen I N-proteinase, Procollagen I/II amino propeptide-processing enzyme, Procollagen N-endopeptidase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1029-1213 of human ADAMTS2 (O95450). PRNISDPSKKSYVVQWLSRPDPDSPIRKISSKGHCQGDKSIFCRMEVLSRYCSIPGYNKLCCKSCNLYNNLTNVEGRIEPPPGKHNDIDVFMPTLPVPTVAMEVRPSPSTPLEVPLNASSTNATEDHPETNAVDEPYKIHGLEDEVQPPNLIPRRPSPYEKTRNQRIQELIDEMRKKEMLGKF | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title