missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAMTS18 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | ADAMTS18 |
|---|---|
| Applications | Immunohistochemistry, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17482333
|
Proteintech
26839-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17472333
|
Proteintech
26839-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The ADAMTS18 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human ADAMTS18. This antibody recognizes human antigen. The ADAMTS18 antibody has been validated for the following applications: IHC, ELISA analysis.
Specifications
| ADAMTS18 | |
| Polyclonal | |
| Rabbit | |
| 170692 | |
| Primary |
| Immunohistochemistry, ELISA | |
| Unconjugated | |
| Human | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title