missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAMTS13 Rabbit anti-Human, Mouse, Rat, Clone: 7B2Z8, Novus Biologicals™
Rabbit Monoclonal Antibody
£167.00 - £413.00
Specifications
| Antigen | ADAMTS13 |
|---|---|
| Clone | 7B2Z8 |
| Dilution | Western Blot 1:500 - 1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18310827
|
Novus Biologicals
NBP3-16038-20UL |
20 μg |
£167.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18336506
|
Novus Biologicals
NBP3-16038-100UL |
100 μg |
£413.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ADAMTS13 Monoclonal antibody specifically detects ADAMTS13 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, ImmunofluorescenceSpecifications
| ADAMTS13 | |
| Western Blot 1:500 - 1:1000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| A disintegrin and metalloproteinase with thrombospondin motifs 13, a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif, 13, ADAM metallopeptidase with thrombospondin type 1 motif, 13, ADAM-TS13, ADAMTS-13, DKFZp434C2322, EC 3.4.24.14, EC 3.4.24.82, EC 3.4.24.87, FLJ42993, MGC118899, MGC118900, TTPADAM-TS 13, vWF-cleaving protease, vWF-CPC9orf8, VWFCPvon Willebrand factor-cleaving protease | |
| A synthetic peptide corresponding to a sequence within amino acids 50-150 of human ADAMTS13 (Q76LX8). LSPGAPLKGRPPSPGFQRQRQRQRRAAGGILHLELLVAVGPDVFQAHQEDTERYVLTNLNIGAELLRDPSLGAQFRVHLVKMVILTEPEGAPNITANLTSS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| 7B2Z8 | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cancer, Signal Transduction | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 11093 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title