missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ADAMTS12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568084
This item is not returnable.
View return policy
Description
ADAMTS12 Polyclonal Antibody for Western Blot
This protein is a disintegrin and metalloproteinase with thrombospondin motifs-12 (ADAMTS12), which is a member of the ADAMTS protein family. Members of the family share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS-1) motif. Individual members of this family differ in the number of C-terminal TS-1 motifs, and some have unique C-terminal domains. The enzyme encoded by this gene contains eight TS-1 motifs. It may play roles in pulmonary cells during fetal development or in tumor processes through its proteolytic activity or as a molecule potentially involved in regulation of cell adhesion.
Specifications
| ADAMTS12 | |
| Polyclonal | |
| Unconjugated | |
| ADAMTS12 | |
| a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 12; a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 12; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 12; a disintegrin-like and metalloprotease with thrombospondin type 1 motif, 12; ADAM; ADAM metallopeptidase with thrombospondin type 1 motif 12; ADAM metallopeptidase with thrombospondin type 1 motif, 12; ADAMs; ADAM-TS 12; ADAMTS12; ADAM-TS12; ADAMTS-12; AI605170; metalloendopeptidases; metalloprotease disintegrin 12; PRO4389; UNQ1918; UNQ1918/PRO4389 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 81792 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P58397 | |
| ADAMTS12 | |
| Synthesized peptide derived from human ADAMTS12. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction