missing translation for 'onlineSavingsMsg'
Learn More

ADAMTS10 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18428889 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18428889 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18428889 Supplier Novus Biologicals Supplier No. NBP299535JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ADAMTS10 Polyclonal antibody specifically detects ADAMTS10 in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Host Species Rabbit
Quantity 0.1 mL
Regulatory Status RUO
Gene ID (Entrez) 81794
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.