missing translation for 'onlineSavingsMsg'
Learn More

ADAMTS10 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18416799 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18416799 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18416799 Supplier Novus Biologicals Supplier No. NBP299535AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ADAMTS10 Polyclonal antibody specifically detects ADAMTS10 in Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAMTS10
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias a disintegrin-like and metalloprotease (reprolysin type) with thrombospondintype 1 motif, 10, ADAM metallopeptidase with thrombospondin type 1 motif, 10, ADAM-TS 10, ADAMTS-10, ADAM-TS10A disintegrin and metalloproteinase with thrombospondin motifs 10, EC 3.4.24.-, EC 3.4.24.82, WMS, zinc metalloendopeptidase
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human ADAMTS10
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 81794
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.