missing translation for 'onlineSavingsMsg'
Learn More

Adamantane, 98%

Product Code. 11427936
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11427936 250 g 250g
11417936 50 g 50g
11407936 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11427936 Supplier Thermo Scientific Alfa Aesar Supplier No. A14041.30
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 281-23-2 | C10H16 | 136.238 g/mol

Adamantane is mainly used for the synthesis of medicine to treat influenza, serve as NMDA receptor channel blockers. Also be used for preparation of advanced lubricants, photographic material, surface active agents, catalysts and other products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Adamantane is mainly used for the synthesis of medicine to treat influenza, serve as NMDA receptor channel blockers. Also be used for preparation of advanced lubricants, photographic material, surface active agents, catalysts and other products.

Solubility
Insoluble in water.

Notes
Store in a cool, dry, well-ventilated area. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 281-23-2
Molecular Formula C10H16
Molecular Weight (g/mol) 136.238
MDL Number MFCD00074719
InChI Key ORILYTVJVMAKLC-UHFFFAOYSA-N
Synonym tricyclo 3.3.1.13,7 decane, adamantan, tricyclo 3.3.1.1 3,7 decane, tricyclo 3.3.1.1 decane, unii-pjy633525u, tricyclo 3.3.1.1∼3,7∼ decane, adamantine, 2-adamantyl, adamantan-1-yl
PubChem CID 9238
ChEBI CHEBI:40519
IUPAC Name adamantane
SMILES C1C2CC3CC1CC(C2)C3

Specifications

Melting Point 266°C to 268°C (sealed tube)
Density 1.07
Quantity 250 g
Beilstein 1901173
Merck Index 14,149
Solubility Information Insoluble in water.
Formula Weight 136.24
Percent Purity 98%
Chemical Name or Material Adamantane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.