missing translation for 'onlineSavingsMsg'
Learn More

Adamantane-1-carbonyl chloride, 97%

Product Code. 11327675
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11327675 10 g 10g
11337675 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11327675 Supplier Thermo Scientific Alfa Aesar Supplier No. L02352.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2094-72-6 | C11H15ClO | 198.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2094-72-6
Molecular Formula C11H15ClO
Molecular Weight (g/mol) 198.69
MDL Number MFCD00074724
InChI Key MIBQYWIOHFTKHD-UHFFFAOYSA-N
Synonym 1-adamantanecarbonyl chloride, adamantanecarbonyl chloride, 1-adamantanecarbonylchloride, 1-adamantoyl chloride, adamantoyl chloride, 1-adamantanoic acid chloride, 1-adamantanecarboxylic acid chloride, adamantane-1-carboxylic acid chloride, 1-adamantylcarbonyl chloride, 1-adamantyl carbonyl chloride
PubChem CID 98915
IUPAC Name adamantane-1-carbonyl chloride
SMILES C1C2CC3CC1CC(C2)(C3)C(=O)Cl

Specifications

Melting Point 48°C to 53°C
Boiling Point 135°C to 136°C (10 mmHg)
Quantity 10 g
UN Number UN3261
Beilstein 389960
Sensitivity Moisture sensitive
Formula Weight 198.69
Percent Purity 97%
Chemical Name or Material Adamantane-1-carbonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.