missing translation for 'onlineSavingsMsg'
Learn More

Adamantane-1,3-dicarboxylic acid, 98%

Product Code. 15476548
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15476548

Brand: Thermo Scientific Alfa Aesar H60081.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 39269-10-8 | C12H14O4 | 222.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39269-10-8
Molecular Formula C12H14O4
Molecular Weight (g/mol) 222.24
MDL Number MFCD00167790
InChI Key PAVQGHWQOQZQEH-UHFFFAOYSA-L
Synonym 1,3-adamantanedicarboxylic acid, 1,3-dicarboxyadamantane, 1s,3s,5r,7r-adamantane-1,3-dicarboxylic acid, 1,3-adamantane dicarboxylic acid, tricyclo 3.3.1.1 sup 3,7 decane-1,3-dicarboxylic acid, tricyclo 3.3.1.13,7 decane-1,3-dicarboxylic acid, tricyclo 3.3.1.1∼3,7∼ decane-1,3-dicarboxylic acid, 1r,3s,5s,7s-adamantane-1,3-dicarboxylic acid, 1,3-adamantanedicarboxylicacid
PubChem CID 64339
IUPAC Name adamantane-1,3-dicarboxylic acid
SMILES [O-]C(=O)C12CC3CC(C1)CC(C3)(C2)C([O-])=O

Specifications

Melting Point 276°C to 278°C
Quantity 1 g
Formula Weight 224.25
Percent Purity 98%
Chemical Name or Material Adamantane-1,3-dicarboxylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.