missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ADAM9 Recombinant Rabbit Monoclonal Antibody (2A0V9)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558017
This item is not returnable.
View return policy
Description
Immunogen sequence: AARPGFQQTS HLSSYEIITP WRLTRERREA PRPYSKQVSY VIQAEGKEHI IHLERNKDLL PEDFVVYTYN KEGTLITDHP NIQNHCHYRG YVEGVHNSSI ALSDCFGLRG LLHLENASYG IEPLQNSSHF EHIIYRMDDV YKEPLKCGVS NKDIEKETAK DEEEEPPSMT QLLRRRRAVL PQ.
ADAM9 is a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. This protein interacts with SH3 domain-containing proteins, binds mitotic arrest deficient 2 beta protein, and is also involved in TPA-induced ectodomain shedding of membrane-anchored heparin-binding EGF-like growth factor.
Specifications
| ADAM9 | |
| Recombinant Monoclonal | |
| 2.51 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| Q13443 | |
| ADAM9 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 29-210 of human ADAM9 (NP_003807.1). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 2A0V9 | |
| Unconjugated | |
| ADAM9 | |
| a disintegrin and metallopeptidase domain 9 (meltrin gamma); a disintegrin and metalloprotease domain 9; a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 9 (meltrin gamma); ADAM; ADAM 9; ADAM metallopeptidase domain 9; ADAM metallopeptidase domain 9 (meltrin gamma); adam9; ADAMs; AU020942; Cellular disintegrin-related protein; cone rod dystrophy 9; CORD9; Disintegrin and metalloproteinase domain-containing protein 9; Kiaa0021; MCMP; MDC 9; MDC9; meltrin gamma; meltrin-gamma; metalloendopeptidases; Metalloprotease/disintegrin/cysteine-rich protein 9; mKIAA0021; Mltng; Myeloma cell metalloproteinase; OTTHUMP00000216481; OTTHUMP00000216484; OTTHUMP00000216485 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 8754 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction