Learn More
Invitrogen™ ADAM9 Human ELISA Kit
Sandwich ELISA Kit
£544.00 - £4280.00
Specifications
Accession Number | Q13443 |
---|---|
Assay Sensitivity | 45 pg/mL |
Assay | Solid-phase sandwich ELISA |
Conjugate | Biotin |
Product Type | ELISA Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15404893
|
Invitrogen™
EHADAM9 |
96 Tests |
£544.00
1 set |
Estimated Shipment: 28-05-2024
Log in to see stock. |
|||||
17884170
|
Invitrogen™
EHADAM9X10 |
10 x 96 Tests |
£4280.00
Pack of 10 |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
17155339
|
Invitrogen™
EHADAM9X5 |
5 x 96 Tests |
£2320.00
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
Description
The Human ADAM9 ELISA quantitates Hu ADAM9 in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu ADAM9. Principle of the method The Human ADAM9 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
The protein encoded by this gene interacts with SH3 domain-containing proteins, binds mitotic arrest deficient 2 beta protein, and is also involved in TPA-induced ectodomain shedding of membrane-anchored heparin-binding EGF-like growth factor. Several alternatively spliced transcript variants have been identified for this gene.Specifications
Q13443 | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
Human | |
Colorimetric Microplate Reader | |
CORD9,MCMP,MDC9,Mltng | |
<10% | |
RUO | |
2°C to 8°C | |
Human | |
4 hr. 45 min. |
45 pg/mL | |
Biotin | |
Growth Factors & Receptors | |
This ELISA antibody pair detects human ADAM9. Other species not determined yet. | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
8754 | |
<12% | |
HRP | |
Extracellular Matrix & Cell Adhesion | |
CORD9, MCMP, MDC9, Mltng | |
1 hr. 20 min. |