missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAM9 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-32260CL1
This item is not returnable.
View return policy
Description
ADAM9 Polyclonal antibody specifically detects ADAM9 in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| ADAM9 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunoprecipitation, Western Blot | |
| CoraFluor 1 | |
| a disintegrin and metalloproteinase domain 9 (meltrin gamma), ADAM 9, ADAM metallopeptidase domain 9, ADAM metallopeptidase domain 9 (meltrin gamma), Cellular disintegrin-related protein, cone rod dystrophy 9, CORD9, disintegrin and metalloproteinase domain-containing protein 9, EC 3.4.24, EC 3.4.24.-, MCMPMDC9KIAA0021Mltng, Meltrin-gamma, Metalloprotease/disintegrin/cysteine-rich protein 9, MLTNG, Myeloma cell metalloproteinase | |
| The immunogen this antibody was made to, maps to a region between residue 400 to 450 of human ADAM Metallopeptidase Domain 9 using the numbering given in entry NP_003807.1 (GeneID 8754). | |
| 0.1 mL | |
| Signal Transduction | |
| 8754 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction