missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAM8 Polyclonal Antibody
ADAM8 Polyclonal Antibody
Brand: Invitrogen PA587946
This item is not returnable.
View return policy
Description
ADAM8 Polyclonal Antibody for Western Blot
Specifications
| ADAM8 | |
| Polyclonal | |
| Unconjugated | |
| ADAM8 | |
| a disintegrin and metallopeptidase domain 8; a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 8; ADAM; ADAM 8; ADAM metallopeptidase domain 8; Adam8; ADAMs; CD156; CD156a; Cell surface antigen MS2; disintegrin and metalloproteinase domain-containing protein 8; E430039A18Rik; human leukocyte differentiation antigen; Macrophage cysteine-rich glycoprotein; metalloendopeptidases; MGC134985; MS2; RGD1566394 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 101, 11501, 499285 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.336 mg/mL | |
| PBS with 50% glycerol and 0.09% sodium azide; pH 7.3 | |
| P78325, Q05910 | |
| ADAM8 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 17-300 of human ADAM8 (NP_0011003). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction