missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ADAM33 Monoclonal Antibody (219515)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA523936
This item is not returnable.
View return policy
Description
In Western blots, no cross-reactivity with recombinant mouse ADAM9, 10, 15, recombinant human (rh) ADAM8, rhTACE/ADAM17, rhADAM19, or rhADAM33 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. This protein is a type I transmembrane protein implicated in asthma and bronchial hyperresponsiveness. Alternative splicing of this gene results in two transcript variants encoding different isoforms.
Specifications
| ADAM33 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q923W9 | |
| Adam33 | |
| Mouse myeloma cell line NS0-derived recombinant mouse ADAM33 Ectodomain Glu205-Ala702. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2b |
| Immunoprecipitation, Western Blot | |
| 219515 | |
| Unconjugated | |
| Adam33 | |
| a disintegrin and metallopeptidase domain 33; a disintegrin and metalloprotease 33; a disintegrin and metalloprotease domain 33; a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 33; ADAM; ADAM 33; ADAM metallopeptidase domain 33; ADAM33; Adaml; ADAMs; C20orf153; disintegrin and metalloproteinase domain-containing protein 33; disintegrin and reprolysin metalloproteinase family protein; DJ964F7.1; DKFZp434K0521; FLJ35308; FLJ36751; HGNC:15478; metalloendopeptidases; MGC149823; MGC71889; RP23-228H14.4; RP5-964F7.2; UNQ873/PRO1891 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 110751 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction