missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ADAM32 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA583709
This item is not returnable.
View return policy
Description
Immunogen sequence: DYKLPRTVPD PLAVKNGSQC DIGRVCVNRE CVESRIIKAS AHVCSQQCSG HGVCDSRNKC HCSPGYKPPN CQIRSKGFSI FPEEDMGSIM E Highest antigen sequence indentity to the following orthologs: Rat - 40%, Mouse - 37%.
This gene encodes a member of the disintegrin family of membrane-anchored proteins that play a role in diverse biological processes such as brain development, fertilization, tumor development and inflammation. This gene is predominantly expressed in the testis. The encoded protein undergoes proteolytic processing to generate a mature polypeptide comprised of an metalloprotease, disintegrin and epidermal growth factor-like domains. This gene is located in a cluster of other disintegrin and metallopeptidase family genes on chromosome 8. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq, Sep 2015].
Specifications
| ADAM32 | |
| Polyclonal | |
| Unconjugated | |
| ADAM32 | |
| a disintegrin and metallopeptidase domain 32; a disintegrin and metalloprotease domain 32; a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 32; ADAM; ADAM 32; ADAM metallopeptidase domain 32; Adam32; ADAMs; Disintegrin and metalloproteinase domain-containing protein 32; FLJ26299; FLJ29004; metalloendopeptidases; metalloprotease/disintegrin; metalloproteinase 12-like protein; testicular tissue protein Li 13; UNQ5982/PRO21340 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 203102 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.10 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TC27 | |
| ADAM32 | |
| Recombinant protein corresponding to Human ADAM32. Recombinant protein control fragment (Product #RP-100296). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction