missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAM30 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA543393
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: RLLLPRHLRV FSFTEHGELL EDHPYIPKDC NYMGSVKESL DSKATISTCM Sequence homology: Cow: 85%; Dog: 77%; Human: 100%
ADAM30 Polyclonal antibody specifically detects ADAM30 in Human samples. It is validated for Western Blot.Specifications
ADAM30 | |
Polyclonal | |
Unconjugated | |
ADAM30 | |
a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 30; ADAM; ADAM 30; ADAM metallopeptidase domain 30; ADAM30; ADAMs; Disintegrin and metalloproteinase domain-containing protein 30; metalloendopeptidases; svph4; UNQ2509/PRO5997 | |
Rabbit | |
Protein A | |
RUO | |
11085 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9UKF2 | |
ADAM30 | |
Synthetic peptide directed towards the N-terminal of human ADAM30 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |