missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ADAM30 Monoclonal Antibody (3E5)
GREENER_CHOICE

Product Code. 15548972
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15548972 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15548972 Supplier Invitrogen™ Supplier No. MA521251

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Peptide Sequence: YKHPKYLELI LLFDQSRYRF VNNNLSQVIH DAILLTGIMD TYFQDVRMRI HLKALEVWTD FNKIRVGYPE LAEVLGRFVI YKKSVLNARL SSDWAHLYLQ.

This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. This gene is testis-specific and contains a polymorphic region, resulting in isoforms with varying numbers of C-terminal repeats.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAM30
Applications ELISA
Classification Monoclonal
Clone 3E5
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene ADAM30
Gene Accession No. Q9UKF2
Gene Alias a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 30; ADAM; ADAM 30; ADAM metallopeptidase domain 30; ADAM30; ADAMs; Disintegrin and metalloproteinase domain-containing protein 30; metalloendopeptidases; svph4; UNQ2509/PRO5997
Gene Symbols ADAM30
Host Species Mouse
Immunogen ADAM30 (NP_068566, 199 a.a. ~ 298 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11085
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.