missing translation for 'onlineSavingsMsg'
Learn More

ADAM30 Antibody (3E5), Novus Biologicals™

Product Code. 18355279 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18355279 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18355279 Supplier Novus Biologicals Supplier No. H00011085M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ADAM30 Monoclonal antibody specifically detects ADAM30 in Human samples. It is validated for ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAM30
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 3E5
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_068566
Gene Alias a disintegrin and metalloproteinase domain 30, ADAM 30, ADAM metallopeptidase domain 30, disintegrin and metalloproteinase domain-containing protein 30, EC 3.4.24.-, svph4
Host Species Mouse
Immunogen ADAM30 (NP_068566, 199 a.a. ~ 298 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. YKHPKYLELILLFDQSRYRFVNNNLSQVIHDAILLTGIMDTYFQDVRMRIHLKALEVWTDFNKIRVGYPELAEVLGRFVIYKKSVLNARLSSDWAHLYLQ
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11085
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.