missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAM29 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81697H
This item is not returnable.
View return policy
Description
ADAM29 Polyclonal antibody specifically detects ADAM29 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| ADAM29 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 11086 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| HRP | |
| ADAM 29, ADAM metallopeptidase domain 29, Cancer/testis antigen 73, CT73a disintegrin and metalloproteinase domain 29, disintegrin and metalloproteinase domain-containing protein 29, EC 3.4.24, EC 3.4.24.58, svph1 | |
| ADAM29 antibody was raised against a 19 amino acid peptide near the carboxy terminus of human ADAM29. The immunogen is located within amino acids 710 - 760 of ADAM29. Amino Acid Squence: EEEKIQRRPHELPPQSQPW | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction