missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAM28 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £386.00
Specifications
| Antigen | ADAM28 |
|---|---|
| Concentration | 1.0mg/mL |
| Dilution | Western Blot 2μg/ml, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 10μg/ml, Immunohistochemistry-Paraffin 1:200 |
| Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18172034
|
Novus Biologicals
NBP2-67246SS |
0.025 mg |
£145.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18160034
|
Novus Biologicals
NBP2-67246 |
0.1 mg |
£386.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ADAM28 Polyclonal specifically detects ADAM28 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| ADAM28 | |
| Western Blot 2μg/ml, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 10μg/ml, Immunohistochemistry-Paraffin 1:200 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| a disintegrin and metalloproteinase domain 28, ADAM metallopeptidase domain 28, ADAM23MDC-L, disintegrin and metalloproteinase domain-containing protein 28, EC 3.4.24, EC 3.4.24.-, eMDC II, eMDCII, Epididymial metalloproteinase-like, disintegrin-like, and cysteine-rich proteinII, MDCLADAM 28, MDC-Lm, MDC-Ls, Metalloproteinase-like, disintegrin-like, and cysteine-rich protein L, metalloproteinase-like, disintegrin-like, and cysteine-rich protein-L | |
| ADAM28 | |
| IgG | |
| Affinity Purified |
| 1.0mg/mL | |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Cancer, Signal Transduction | |
| PBS with 0.02% Sodium Azide | |
| 10863 | |
| Partial recombinant human ADAM28 protein (amino acids 72-623) [UniProt Q9UKQ2] | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title