missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ADAM22 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565610
This item is not returnable.
View return policy
Description
Immunogen sequence: LSPAKSPSSST GSIASSRKYP YPMPPLPDED KKVNRQSARL WETSI Highest antigen sequence identity to the following orthologs - mouse 91%, rat 96%.
This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biological processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. This gene is highly expressed in the brain and may function as an integrin ligand in the brain. Alternative splicing results in several transcript variants.
Specifications
| ADAM22 | |
| Polyclonal | |
| Unconjugated | |
| Adam22 | |
| 2900022I03Rik; a disintegrin and metallopeptidase domain 22; a disintegrin and metalloprotease domain (ADAM) 22; a disintegrin and metalloprotease domain 22; a disintegrin and metalloproteinase; a disintegrin and metalloproteinase domain 22; ADAM; ADAM 22; ADAM metallopeptidase domain 22; Adam22; ADAMs; AI854032; Disintegrin and metalloproteinase domain-containing protein 22; MDC2; metalloendopeptidases; Metalloproteinase-disintegrin ADAM22-3; metalloproteinase-like, disintegrin-like, and cysteine-rich protein 2; MGC149832 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 53616 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9P0K1 | |
| Adam22 | |
| Recombinant Human ADAM22. Recombinant protein control fragment (Product #RP-106275). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction