missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAM22 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £484.00
Specifications
| Antigen | ADAM22 |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18236411
|
Novus Biologicals
NBP2-58310 |
100 μL |
£484.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18683688
|
Novus Biologicals
NBP2-58310-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ADAM22 Polyclonal specifically detects ADAM22 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| ADAM22 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 53616 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:LSPAKSPSSSTGSIASSRKYPYPMPPLPDEDKKVNRQSARLWETSI | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| ADAM 22, ADAM metallopeptidase domain 22, metalloproteinase-disintegrin ADAM22-3, metalloproteinase-like, disintegrin-like, and cysteine-rich protein 2, MGC149832 | |
| ADAM22 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title