missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ADAM21 293T Cell Transient Overexpression Lysate
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.01mL
Specifications
Specifications
| Quantity | 0.1 mL |
| Content And Storage | Store at -80°C. Avoid freeze-thaw cycles. |
| Target | ADAM21 |
| Synonym | a disintegrin and metalloproteinase domain 21, ADAM 21, ADAM metallopeptidase domain 21, ADAM metallopeptidase domain 21, preproprotein, ADAM31, disintegrin and metalloproteinase domain-containing protein 21, EC 3.4.24.-, MGC125389 |
| Gene Symbol | ADAM21 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction