missing translation for 'onlineSavingsMsg'
Learn More

ADAM2 Antibody (4A2), Novus Biologicals™

Product Code. 18328877 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18328877 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18328877 Supplier Novus Biologicals Supplier No. H00002515M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ADAM2 Monoclonal antibody specifically detects ADAM2 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen ADAM2
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 4A2
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_001455
Gene Alias ADAM metallopeptidase domain 2, Cancer/testis antigen 15, CRYN2, CT15ADAM 2, disintegrin and metalloproteinase domain-containing protein 2, EC 3.4.24, fertilin beta, Fertilin subunit beta, FTNBCRYN1, PH-30, PH-30b, PH30PH30-beta
Host Species Mouse
Immunogen ADAM2 (NP_001455, 376 a.a. ~ 475 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. RLDPFFKQQAVCGNAKLEAGEECDCGTEQDCALIGETCCDIATCRFKAGSNCAEGPCCENCLFMSKERMCRPSFEECDLPEYCNGSSASCPENHYVQTGH
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2515
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.