missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ADAM17 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20259-1-AP-20UL
This item is not returnable.
View return policy
Description
This antibody is specific to ADAM17.
This gene encodes a member of the ADAM (a disintegrin and metalloprotease domain) family. Members of this family are membrane-anchored proteins structurally related to snake venom disintegrins, and have been implicated in a variety of biologic processes involving cell-cell and cell-matrix interactions, including fertilization, muscle development, and neurogenesis. The protein encoded by this gene functions as a tumor necrosis factor-alpha converting enzyme; binds mitotic arrest deficient 2 protein; and also plays a prominent role in the activation of the Notch signaling pathway.Specifications
| ADAM17 | |
| Polyclonal | |
| Unconjugated | |
| ADAM17 | |
| ADAM 17, ADAM17, CD156b, cSVP, Snake venom like protease, TACE, TNF alpha convertase, TNF alpha converting enzyme | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 6868 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P78536 | |
| ADAM17 | |
| Peptide | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction